This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04721502_m1
See other BCAP31 GE Assays ›
Gene Symbol:
BCAP31
Gene Name:
B cell receptor associated protein 31
Gene Aliases:
-
Chromosome Location:
Chr.Un|NW_017189840: 75802 - 104943 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04721502_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCAP31
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018044779.1 XP_017900268.1 4 - 5 492 63
XM_018044780.1 XP_017900269.1 4 - 5 470 63
GenBank mRNA JO352584.2 - 4 - 5 625 63
JO502968.2 - 4 - 5 655 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0