This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04718595_m1
See other LOC102184834 GE Assays ›
Gene Symbol:
LOC102184834
Gene Name:
voltage-dependent R-type calcium channel subunit alpha-1E
Gene Aliases:
CACNA1E
Chromosome Location:
Chr.16: 61218598 - 61557291 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04718595_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102184834
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018060680.1 XP_017916169.1 43 - 44 5636 79
XM_018060681.1 XP_017916170.1 42 - 43 5579 79
XM_018060682.1 XP_017916171.1 44 - 45 6940 79

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5c796d7db4-nk9d7:80/100.66.128.142:80.
git-commit: d3b3aa98da0ae1b146c174fddb0766cfa6979daf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0