Assay Details
Gene Symbol: GRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048796.1 | XP_017904285.1 | 6 - 7 | 891 | 60 | |
GenBank mRNA | JO599599.2 | - | 2 - 3 | 2559 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
