This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04712640_m1
See other VSIG10 GE Assays ›
Gene Symbol:
VSIG10
Gene Name:
V-set and immunoglobulin domain containing 10
Gene Aliases:
-
Chromosome Location:
Chr.17: 14066149 - 14105985 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04712640_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005691462.3 XP_005691519.1 5 - 6 1463 68
XM_013970608.2 XP_013826062.1 5 - 6 1469 68
XM_013970609.2 XP_013826063.1 5 - 6 1473 68
GenBank mRNA KA323535.1 - 4 - 5 1349 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0