This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04711222_m1
See other JAM2 GE Assays ›
Gene Symbol:
JAM2
Gene Name:
junctional adhesion molecule 2
Gene Aliases:
-
Chromosome Location:
Chr.1: 9118653 - 9199087 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04711222_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: JAM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005674736.2 XP_005674793.1 4 - 5 647 68
XM_018051483.1 XP_017906972.1 4 - 5 432 68
GenBank mRNA JO391161.2 - 3 - 4 380 68
JO449810.2 - 2 - 3 265 68
JO606363.2 - 4 - 5 449 68
JO635932.2 - 2 - 3 192 68
JT706256.1 - 7 - 8 485 68
JT738174.1 - 9 - 10 1131 68
KA323898.1 - 6 - 7 1827 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0