Assay Details
Gene Symbol: NECTIN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005677180.3 | XP_005677237.1 | 8 - 9 | 1964 | 82 | |
XM_005677181.3 | XP_005677238.1 | 8 - 9 | 1960 | 82 | ||
XM_005677184.3 | XP_005677241.1 | 8 - 9 | 1598 | 82 | ||
XM_005677185.3 | XP_005677242.1 | 8 - 9 | 1595 | 82 | ||
GenBank mRNA | KA319874.1 | - | 8 - 9 | 1585 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
