Assay Details
Gene Symbol: LOC102188839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043672.1 | XP_017899161.1 | 8 - 9 | 1094 | 77 | |
GenBank mRNA | JO623579.2 | - | 1894 | 77 | ||
JT662416.1 | - | 1730 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
