This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04704919_m1
See other LOC102188839 GE Assays ›
Gene Symbol:
LOC102188839 GNL3L
Gene Name:
guanine nucleotide-binding protein-like 3-like protein
G protein nucleolar 3 like
Gene Aliases:
-
-
Chromosome Location:
Chr.X|NW_017189516: 21846971 - 21860943 on Build ARS1
Chromosome Location:
Chr.X|NW_017189516: 24144414 - 24160635 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
-
Assay ID Ch04704919_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102188839
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018043672.1 XP_017899161.1 4 - 5 467 68
GenBank mRNA JO623579.2 - 1267 68
Gene Symbol: GNL3L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA JO623580.2 - 1267 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7cdd86b58d-kqgnb:80/100.66.128.142:80.
git-commit: 00e1f9c870be1646ae16b59a7cca7819e45b49ab
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0