Assay Details
Gene Symbol: DNASE1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018040615.1 | XP_017896104.1 | 3 - 4 | 399 | 67 | |
GenBank mRNA | KA312513.1 | - | 6 - 7 | 3781 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
