This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04692791_m1
See other MRPL58 GE Assays ›
Gene Symbol:
MRPL58
Gene Name:
mitochondrial ribosomal protein L58
Gene Aliases:
-
Chromosome Location:
Chr.19: 55633835 - 55640862 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04692791_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MRPL58
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018063840.1 XP_017919329.1 2 - 3 264 105
XM_018063841.1 XP_017919330.1 2 - 3 172 105
GenBank mRNA JO577132.1 - 4 - 5 700 105
JO577133.1 - 4 - 5 700 105

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

translation factor translation release factor translational protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0