Assay Details
Gene Symbol: BABAM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005686868.3 | XP_005686925.1 | 11 - 12 | 1342 | 70 | |
GenBank mRNA | JO431565.2 | - | 11 - 12 | 1228 | 70 | |
KA340956.1 | - | 11 - 12 | 1237 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
