This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04692049_m1
See other CEBPZOS GE Assays ›
Gene Symbol:
CEBPZOS LOC108633904
Gene Name:
CEBPZ opposite strand
protein CEBPZOS-like
Gene Aliases:
-
-
Chromosome Location:
Chr.11: 19485867 - 19492413 on Build ARS1
Chromosome Location:
Chr.25: 32479949 - 32480921 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
-
Assay ID Ch04692049_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEBPZOS
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005686493.3 XP_005686550.1 3 - 4 320 96
XM_013967556.2 XP_013823010.1 2 - 3 187 96
XM_018055127.1 XP_017910616.1 3 - 4 249 96
XM_018055128.1 XP_017910617.1 3 - 4 293 96
XM_018055129.1 XP_017910618.1 2 - 3 209 96
GenBank mRNA KA350448.1 - 3 - 4 311 96
Gene Symbol: LOC108633904
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018040888.1 XP_017896377.1 1 629 96

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0