Assay Details
Gene Symbol: C2CD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038504.1 | XP_017893993.1 | 8 - 9 | 858 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Gene Symbol: C2CD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038504.1 | XP_017893993.1 | 8 - 9 | 858 | 67 |
Set Membership: |
![]() ![]() ![]() ![]() |