Assay Details
Gene Symbol: TIMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005692403.3 | XP_005692460.1 | 4 - 5 | 435 | 59 | |
XM_018062393.1 | XP_017917882.1 | 5 - 6 | 368 | 59 | ||
XM_018062394.1 | XP_017917883.1 | 4 - 5 | 359 | 59 | ||
XM_018062395.1 | XP_017917884.1 | 4 - 5 | 438 | 59 | ||
GenBank mRNA | KA312999.1 | - | 4 - 5 | 420 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
