This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04677186_m1
See other LGALS3BP GE Assays ›
Gene Symbol:
LGALS3BP
Gene Name:
galectin 3 binding protein
Gene Aliases:
-
Chromosome Location:
Chr.19: 52624713 - 52634207 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04677186_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LGALS3BP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018063671.1 XP_017919160.1 4 - 5 547 63
XM_018063672.1 XP_017919161.1 4 - 5 547 63
GenBank mRNA JO623860.2 - 4 - 5 538 63
KA338334.1 - 2 - 3 1481 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-fd98957d6-rbs6c:80/100.66.128.142:80.
git-commit: 278a916a254c4ca07f9ccbb2b9efa8cd7578b5be
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0