This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04675249_m1
See other LOC102181879 GE Assays ›
Gene Symbol:
LOC102181879 DSP
Gene Name:
desmoplakin
desmoplakin
Gene Aliases:
-
-
Chromosome Location:
Chr.Un|NW_017206906: 47 - 6656 on Build ARS1
Chromosome Location:
Chr.23: 2528372 - 2572188 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
-
Assay ID Ch04675249_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102181879
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018045406.1 XP_017900895.1 3943 107
Gene Symbol: DSP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018038784.1 XP_017894273.1 21 - 22 3256 107
XM_018038785.1 XP_017894274.1 21 - 22 3135 107

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

intermediate filament binding protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-7c5f86c6b-2xgzq:80/100.66.128.142:80.
git-commit: c124ca8d7fcef91bd439783ab4009544f5b84403
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0