Assay Details
Gene Symbol: C3H1orf109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013962984.2 | XP_013818438.1 | 3 - 4 | 717 | 69 | |
XM_013962986.2 | XP_013818440.1 | 3 - 4 | 717 | 69 | ||
GenBank mRNA | KA305759.1 | - | 2 - 3 | 815 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
