This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04672594_m1
See other IL1R2 GE Assays ›
Gene Symbol:
IL1R2
Gene Name:
interleukin 1 receptor type 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 6611731 - 6648491 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04672594_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL1R2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005686291.3 XP_005686348.3 6 - 7 821 67
XM_005686292.3 XP_005686349.2 6 - 7 906 67
GenBank mRNA KA330115.1 - 3 - 4 1012 67

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0