This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04671357_m1
See other GATA6 GE Assays ›
Gene Symbol:
GATA6
Gene Name:
GATA binding protein 6
Gene Aliases:
-
Chromosome Location:
Chr.24: 34498573 - 34532807 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04671357_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GATA6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018039631.1 XP_017895120.1 4 - 5 1722 84
XM_018039632.1 XP_017895121.1 6 - 7 3208 84
XM_018039633.1 XP_017895122.1 4 - 5 2147 84
XM_018039634.1 XP_017895123.1 4 - 5 1757 84
GenBank mRNA JO394638.2 - 929 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56ffd697dc-85kz4:80/100.66.135.66:80.
git-commit: 082a6d415be982b1f92c35e55dd71609d4576d3e
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.1-2025.09.37-1.0