This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04667001_m1
See other SCN4B GE Assays ›
Gene Symbol:
SCN4B
Gene Name:
sodium voltage-gated channel beta subunit 4
Gene Aliases:
-
Chromosome Location:
Chr.15: 53493099 - 53514072 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04667001_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SCN4B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018059727.1 XP_017915216.1 3 - 4 882 62
GenBank mRNA JO633012.2 - 3 - 4 857 62
JT743221.1 - 3 - 4 674 62

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

myelin protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-56696975f8-skfwn:80/100.66.128.142:80.
git-commit: 795bdb1f794b4b81290d0c522fee4b91276a707f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0