This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04664832_m1
See other PPP2R2B GE Assays ›
Gene Symbol:
PPP2R2B
Gene Name:
protein phosphatase 2 regulatory subunit Bbeta
Gene Aliases:
-
Chromosome Location:
Chr.7: 52197856 - 52676281 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04664832_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PPP2R2B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005683107.3 XP_005683164.1 7 - 8 1371 66
XM_005683109.3 XP_005683166.1 8 - 9 1903 66
XM_013965670.2 XP_013821124.1 4 - 5 590 66
XM_018050189.1 XP_017905678.1 8 - 9 1565 66
XM_018050190.1 XP_017905679.1 8 - 9 1302 66

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0