This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04664465_g1
See other AGR2 GE Assays ›
Gene Symbol:
AGR2
Gene Name:
anterior gradient 2, protein disulphide isomerase family member
Gene Aliases:
-
Chromosome Location:
Chr.4: 95221711 - 95234987 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04664465_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005678997.3 XP_005679054.1 6 - 7 575 73
XM_005678998.3 XP_005679055.1 6 - 7 553 73
XM_013963137.2 XP_013818591.1 6 - 7 536 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

reductase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0