Assay Details
Gene Symbol: SLC2A4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314227.1 | NP_001301156.1 | 4 - 5 | 589 | 61 | |
GenBank mRNA | JO350513.2 | - | 7 - 8 | 2535 | 61 | |
JQ343218.1 | - | 4 - 5 | 589 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
