This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04654000_m1
See other LDH-A GE Assays ›
Gene Symbol:
LDH-A
Gene Name:
lactate dehydrogenase
Gene Aliases:
LDHA
Chromosome Location:
Chr.29: 25683920 - 25695934 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04654000_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LDH-A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285718.1 NP_001272647.1 1 135 68
XM_013975786.2 XP_013831240.2 2 - 3 390 68
XM_018042654.1 XP_017898143.1 2 - 3 367 68
GenBank mRNA GU144018.1 - 1 135 68
JO580468.1 - 2 - 3 327 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Both primers and probe map within a single exon

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-69fc894774-tvlwx:80/100.66.128.142:80.
git-commit: 8e82defa9bf25b3b77800b5e306013d4ba941391
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0