Assay Details
Gene Symbol: CIDEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001314221.1 | NP_001301150.1 | 1 - 2 | 131 | 75 | |
XM_018039339.1 | XP_017894828.1 | 2 - 3 | 248 | 75 | ||
XM_018039340.1 | XP_017894829.1 | 2 - 3 | 248 | 75 | ||
GenBank mRNA | JO624748.2 | - | 3 - 4 | 614 | 75 | |
KP120985.1 | - | 1 - 2 | 131 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
