This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04653796_g1
See other AGPAT1 GE Assays ›
Gene Symbol:
AGPAT1
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 1
Gene Aliases:
LPAAT
Chromosome Location:
Chr.23: 22730563 - 22739667 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04653796_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AGPAT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285761.1 NP_001272690.1 4 - 5 617 58
XM_013973951.2 XP_013829405.1 3 - 4 1247 58
XM_013973952.2 XP_013829406.1 4 - 5 778 58
XM_018038493.1 XP_017893982.1 4 - 5 3069 58
XM_018038494.1 XP_017893983.1 4 - 5 2955 58
XM_018038495.1 XP_017893984.1 4 - 5 2912 58
GenBank mRNA HM623913.1 - 4 - 5 617 58
JO578414.1 - 4 - 5 817 58
JT674928.1 - 5 - 6 936 58

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

acyltransferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-67c6c7f55f-2g6mz:80/100.66.128.142:80.
git-commit: 92b42c745e7842b39e6d37af9c4064bd81a92e4d
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/search_redesign_header