Assay Details
Gene Symbol: MMADHC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285614.1 | NP_001272543.1 | 3 - 4 | 474 | 63 | |
XM_013967881.2 | XP_013823335.1 | 5 - 6 | 666 | 63 | ||
GenBank mRNA | EF151288.1 | - | 3 - 4 | 474 | 63 | |
KA307491.1 | - | 5 - 6 | 660 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
