This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04313505_g1
See other FGF11 GE Assays ›
Gene Symbol:
FGF11
Gene Name:
fibroblast growth factor 11
Gene Aliases:
-
Chromosome Location:
Chr.19: 26926588 - 26933626 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04313505_g1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018064579.1 XP_017920068.1 3 - 4 2145 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0