This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04305102_g1
See other CALM2 GE Assays ›
Gene Symbol:
CALM2
Gene Name:
calmodulin 2
Gene Aliases:
-
Chromosome Location:
Chr.11: 29156106 - 29169683 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04305102_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CALM2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_005686574.3 XP_005686631.1 5 - 6 593 77
XM_018055199.1 XP_017910688.1 5 - 6 525 77
GenBank mRNA JO503857.2 - 5 - 6 506 77
JP350598.2 - 622 77

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

calmodulin-related

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0