Assay Details
Gene Symbol: B4GALNT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018048652.1 | XP_017904141.1 | 4 - 5 | 467 | 57 | |
XM_018048653.1 | XP_017904142.1 | 4 - 5 | 467 | 57 | ||
GenBank mRNA | KA305094.1 | - | 2056 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
