This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Ch04299580_g1
See other DVL1 GE Assays ›
Gene Symbol:
DVL1
Gene Name:
dishevelled segment polarity protein 1
Gene Aliases:
-
Chromosome Location:
Chr.16: 49681973 - 49693800 on Build ARS1
Species:
Goat
Species Specific ID (Flybase ID):
-
Assay ID Ch04299580_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DVL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_018060533.1 XP_017916022.1 10 - 11 1141 61
XM_018060534.1 XP_017916023.1 10 - 11 1138 61
XM_018060535.1 XP_017916024.1 10 - 11 1123 61
XM_018060536.1 XP_017916025.1 10 - 11 1120 61
XM_018060537.1 XP_017916026.1 10 - 11 1141 61
GenBank mRNA JO412552.2 - 591 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Capra hircus Probe spans exons

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0