Assay Details
Gene Symbol: ADGRF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005696388.3 | XP_005696445.2 | 4 - 5 | 507 | 66 | |
XM_005696389.3 | XP_005696446.2 | 5 - 6 | 605 | 66 | ||
GenBank mRNA | KA309705.1 | - | 2 - 3 | 216 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
