Assay Details
Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060097.1 | XP_017915586.1 | 2 - 3 | 369 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Gene Symbol: DYRK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018060097.1 | XP_017915586.1 | 2 - 3 | 369 | 70 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |