Assay Details
Gene Symbol: PRR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018038845.1 | XP_017894334.1 | 2 - 3 | 538 | 89 | |
GenBank mRNA | JO604118.2 | - | 2 - 3 | 198 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
