Assay Details
Gene Symbol: LOC102188839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043672.1 | XP_017899161.1 | 2 - 3 | 207 | 62 | |
GenBank mRNA | JO623579.2 | - | 1007 | 62 |
Gene Symbol: GNL3L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | JO623580.2 | - | 1007 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
