Assay Details
Gene Symbol: LOC102188839 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_018043672.1 | XP_017899161.1 | 7 - 8 | 870 | 85 | |
GenBank mRNA | JO623579.2 | - | 1670 | 85 | ||
JT662416.1 | - | 1506 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
