This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04633474_gH
See other LOC100771856 GE Assays ›
Gene Symbol:
LOC100771856
Gene Name:
core histone macro-H2A.1
Gene Aliases:
H2afy, I79_003878
Chromosome Location:
Chr.Un|NW_003613681: 133707 - 187255 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04633474_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100771856
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003498154.2 XP_003498202.1 7 - 8 975 61
XM_003498155.2 XP_003498203.1 7 - 8 984 61
XM_007622986.1 XP_007621176.1 7 - 8 996 61
XM_007622992.1 XP_007621182.1 7 - 8 993 61
XM_007622997.1 XP_007621187.1 8 - 9 1004 61
XM_007623015.1 XP_007621205.1 7 - 8 981 61
XM_007623030.1 XP_007621220.1 7 - 8 972 61
GenBank mRNA JI882472.1 - 3 - 4 366 61
JP039114.1 - 3 - 4 360 61

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

histone

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0