Assay Details
Gene Symbol: LOC100771856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498154.2 | XP_003498202.1 | 2 - 3 | 315 | 86 | |
XM_003498155.2 | XP_003498203.1 | 2 - 3 | 315 | 86 | ||
XM_007623015.1 | XP_007621205.1 | 2 - 3 | 315 | 86 | ||
XM_007623030.1 | XP_007621220.1 | 2 - 3 | 315 | 86 | ||
GenBank mRNA | JI882473.1 | - | 2 - 3 | 228 | 86 | |
JP039113.1 | - | 2 - 3 | 228 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
