Assay Details
Gene Symbol: Bmf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007644010.1 | XP_007642200.1 | 2 - 3 | 333 | 66 | |
XM_007644012.1 | XP_007642202.1 | 2 - 3 | 420 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
