This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04628507_m1
See other N6AMT2 GE Assays ›
Gene Symbol:
N6amt2
Gene Name:
N-6 adenine-specific DNA methyltransferase 2 (putative)
Gene Aliases:
I79_006689
Chromosome Location:
Chr.Un|NW_003613790: 1406638 - 1423706 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04628507_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: N6amt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003500427.2 XP_003500475.1 4 - 5 416 69
XM_007641167.1 XP_007639357.1 4 - 5 656 69
XM_007641168.1 XP_007639358.1 4 - 5 540 69
XM_007641169.1 XP_007639359.1 4 - 5 387 69
XM_007641170.1 XP_007639360.1 4 - 5 571 69
XM_007641171.1 XP_007639361.1 3 - 4 281 69
GenBank mRNA JI879708.1 - 4 - 5 368 69

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0