Assay Details
Gene Symbol: LOC100761828 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003515674.2 | XP_003515722.2 | 3 - 4 | 590 | 64 | |
GenBank mRNA | JI896810.1 | - | 3 - 4 | 417 | 64 | |
JP057458.1 | - | 3 - 4 | 417 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
