This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04603948_m1
See other KCNB1 GE Assays ›
Gene Symbol:
Kcnb1
Gene Name:
potassium voltage-gated channel, Shab-related subfamily, member 1
Gene Aliases:
I79_009076
Chromosome Location:
Chr.Un|NW_003613899: 201365 - 284317 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04603948_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Kcnb1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007643099.1 XP_007641289.1 2 - 3 608 74

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Ion Channel 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

voltage-gated ion channel

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0