Assay Details
Gene Symbol: Golga7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643919.1 | XP_007642109.1 | 1 - 2 | 360 | 109 | |
XM_007643920.1 | XP_007642110.1 | 1 - 2 | 360 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
