Assay Details
Gene Symbol: Rpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003504904.2 | XP_003504952.1 | 8 - 9 | 661 | 76 | |
GenBank mRNA | JI895379.1 | - | 8 - 9 | 660 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
