Assay Details
Gene Symbol: Cab39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514280.2 | XP_003514328.1 | 4 - 5 | 605 | 73 | |
GenBank mRNA | JI867632.1 | - | 3 - 4 | 443 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
