Assay Details
Gene Symbol: Gpr56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509803.2 | XP_003509851.1 | 10 - 11 | 1333 | 72 | |
XM_007650448.1 | XP_007648638.1 | 10 - 11 | 1561 | 72 | ||
XM_007650449.1 | XP_007648639.1 | 10 - 11 | 1616 | 72 | ||
XM_007650450.1 | XP_007648640.1 | 10 - 11 | 1405 | 72 | ||
XM_007650451.1 | XP_007648641.1 | 10 - 11 | 1364 | 72 | ||
XM_007650452.1 | XP_007648642.1 | 10 - 11 | 1344 | 72 | ||
XM_007650453.1 | XP_007648643.1 | 10 - 11 | 1546 | 72 | ||
XM_007650454.1 | XP_007648644.1 | 10 - 11 | 1600 | 72 | ||
GenBank mRNA | JI870114.1 | - | 8 - 9 | 1222 | 72 | |
JP062986.1 | - | 9 - 10 | 1309 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
