Assay Details
Gene Symbol: Lrp2bp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509815.2 | XP_003509863.2 | 4 - 5 | 872 | 63 | |
XM_007650462.1 | XP_007648652.1 | 6 - 7 | 622 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
