This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04541555_m1
See other FIGF GE Assays ›
Gene Symbol:
Figf
Gene Name:
c-fos induced growth factor (vascular endothelial growth factor D)
Gene Aliases:
I79_009992
Chromosome Location:
Chr.Un|NW_003613952: 334903 - 370580 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04541555_m1
Size
Availability Made To Order
Catalog # 4351372
Price 374.00
Your Price
Online offer:
374.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Figf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007643841.1 XP_007642031.1 5 - 6 1506 93
XM_007643842.1 XP_007642032.1 5 - 6 1506 93
XM_007643843.1 XP_007642033.1 5 - 6 669 93
XM_007643844.1 XP_007642034.1 5 - 6 1491 93
GenBank mRNA JI873754.1 - 5 - 6 836 93
JP043745.1 - 5 - 6 836 93

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Probe spans exons

Panther Classification:

Molecular Function -

growth factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-4mbwj:80/100.66.130.94:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0