This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04536415_m1
See other VAPA GE Assays ›
Gene Symbol:
Vapa
Gene Name:
VAMP (vesicle-associated membrane protein)-associated protein A, 33kDa
Gene Aliases:
I79_016331
Chromosome Location:
Chr.Un|NW_003614535: 624689 - 639551 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04536415_m1
Best Coverage
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Vapa
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_007649029.1 XP_007647219.1 3 - 4 380 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Inventoried Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein membrane trafficking regulatory protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0