This product has been added to your favorites list. Go to My Favorites

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Cg04525254_m1
See other PLP1 GE Assays ›
Gene Symbol:
Plp1
Gene Name:
proteolipid protein 1
Gene Aliases:
I79_020636
Chromosome Location:
Chr.Un|NW_003615303: 41264 - 139379 on Build CriGri-PICRH-1.0
Species:
Cricetulus griseus
Species Specific ID (Flybase ID):
-
Assay ID Cg04525254_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Plp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_003511986.2 XP_003512034.1 15 - 16 2387 60
XM_007652494.1 XP_007650684.1 15 - 16 2492 60
XM_007652495.1 XP_007650685.1 15 - 16 2500 60
XM_007652496.1 XP_007650686.1 14 - 15 2384 60
XM_007652497.1 XP_007650687.1 13 - 14 2279 60
XM_007652498.1 XP_007650688.1 15 - 16 2489 60
XM_007652499.1 XP_007650689.1 14 - 15 2344 60
XM_007652500.1 XP_007650690.1 14 - 15 2418 60
GenBank mRNA JI888526.1 - 5 - 6 510 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Probe spans exons

Panther Classification:

Molecular Function -

myelin protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-5fc94fc45-hqqk4:80/100.66.128.142:80.
git-commit: 7a6627b71d15189168043ee5c63248cc100e5965
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.0-2025.10.41-1.0